Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1
50 g, 3,0 ± mg
Catalog # 611-8286
Supplier:   VWR Collection
VWR®, Calibration Weights, Class M1
Catalog # 611-8286
Supplier:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1
50 g, 3,0 ± mg
Price
58,20 €
Log in for availability
1 * 1 items
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Delivery information: All weights are supplied in plastic cases.
Specifications
  • Weight:
    50 g
  • Seq:
    15
  • Tolerance:
    3,0 ± mg
  • Cat. No.:
    611-8286