VWR®, Calibration Weights, Class M1
20 mg, 0,3 ± mg
Catalog # 611-8276
Supplier: VWR Collection
Jump to:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.
Specifications
- Weight:20 mg
- Seq:05
- Tolerance:0,3 ± mg