VWR®, Calibration Weights, Class M1
1 mg, 0,2 ± mg
Catalog # 611-8272
Supplier: VWR Collection
Jump to:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Delivery information: All weights are supplied in plastic cases.
Documents
Specifications
- Weight:1 mg
- Seq:01
- Tolerance:0,2 ± mg