VWR®, Calibration Weights, Class M1
1 kg, 50 ± mg
Catalog # 611-8268
Supplier: VWR Collection
Jump to:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.
Specifications
- Weight:1 kg
- Seq:44
- Tolerance:50 ± mg



