Funnels, powder, Nalgene®
79 mm, 18 mm, PP, 30 mm, Powder funnels, 115 ml, 85 mm hieght
221-0091
PPCO, translucent.
- Very good chemical resistance
- Outer ribs prevent air from being trapped
- Autoclavable
For transferring powdery materials. Parallel stem prevents blockages.
This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.
- Funnel O-Ø:79 mm
- Description:Powder funnels, 115 ml, 85 mm hieght
- Material:PP
- Seq:02
- Stem O-Ø:18 mm
- Stem length:30 mm
- Cat. No.:221-0091
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact



