Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1
10 mg, 0,25 ± mg
Catalog # 611-8253
Supplier:   VWR Collection
VWR®, Calibration Weights, Class M1
Catalog # 611-8253
Supplier:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1
10 mg, 0,25 ± mg
Price
13,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Delivery information: All weights are supplied in plastic cases.
Specifications
  • Weight:
    10 mg
  • Seq:
    28
  • Tolerance:
    0,25 ± mg