Orders LinkContactUsLinkComponent
Condensers, Dimroth, DURAN®
Condensers, Dimroth, DURAN®
160 mm, 14/23 NS, 14/23 NS
Catalog # SCOT242546101
Supplier:   DWK Life Sciences
Condensers, Dimroth, DURAN®
Catalog # SCOT242546101
Supplier:  DWK Life Sciences
Condensers, Dimroth, DURAN®
Condensers, Dimroth, DURAN®
160 mm, 14/23 NS, 14/23 NS
Price
85,30 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. Water supplied through plastic connectors.
The Dimroth condenser comprises a coil condenser located within a tube. This condenser type has a larger heat exchange surface and thus a better cooling effect than the Liebig or Allihn condenser.
Specifications
  • Cone size:
    14/23 NS
  • Jacket length:
    160 mm
  • Socket size:
    14/23 NS