Filling and venting closures, Nalgene®
83B, Nalgene bottles and carboys with 83B closures, 6,3 mm
Supplier: Thermo Fisher Scientific
Filling and venting closures, Nalgene®
Catalog # 216-8913
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PP, white with TPE seals and TPE caps.
- Fit all large Nalgene® carboys or bottles with 53 mm (53 B) or 83 mm (83 B) screw caps
- Choice of 6,3 mm or 12,7 mm tube for the 83 mm cap (83 B)
- Autoclavable
Screw closures for the sterile liquid transfer of nutrient media, biological reagents, pure water and chemicals. For applications requiring the use of bioreactors and fermenters.
: Pack includes the cap, Nalgene® platinum-treated silicone tubing for aspirating and dispensing and a splash guard for filling.
Specifications
- Cap Size:83B
- For:Nalgene bottles and carboys with 83B closures
- Tubing int.Ø:6,3 mm
- Seq:02
- Cat. No.:216-8913
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact



