Order Entry
Export
Orders LinkContactUsLinkComponent
 

Filling and venting closures, Nalgene®

83B, Nalgene bottles and carboys with 83B closures, 12,7 mm
Catalog # 216-8912
   Low Manufacturing Impact
Filling and venting closures, Nalgene®
Catalog # 216-8912
   Low Manufacturing Impact
Filling and venting closures, Nalgene®
Filling and venting closures, Nalgene®

83B, Nalgene bottles and carboys with 83B closures, 12,7 mm

Price
76,30 €
Log in for availability
1 * 1 items
Product Details & Documents

PP, white with TPE seals and TPE caps.

  • Fit all large Nalgene® carboys or bottles with 53 mm (53 B) or 83 mm (83 B) screw caps
  • Choice of 6,3 mm or 12,7 mm tube for the 83 mm cap (83 B)
  • Autoclavable

Screw closures for the sterile liquid transfer of nutrient media, biological reagents, pure water and chemicals. For applications requiring the use of bioreactors and fermenters.

 : Pack includes the cap, Nalgene® platinum-treated silicone tubing for aspirating and dispensing and a splash guard for filling.

Specifications
  • Cap Size:
    83B
  • For:
    Nalgene bottles and carboys with 83B closures
  • Tubing int.Ø:
    12,7 mm
  • Seq:
    03
  • Cat. No.:
    216-8912
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact