Order Entry
Export
Orders LinkContactUsLinkComponent
 

Funnels with long stem, DURAN®

150 mm, 6 mm, 80 mm
  221-1181
Funnels with long stem, DURAN®
  221-1181
Funnels with long stem, DURAN®
Funnels with long stem, DURAN®

150 mm, 6 mm, 80 mm

 
35,40 €
   
1 * 1 items
 

DURAN®, borosilicate glass 3,3, clear.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

With smooth internal walls. Cone angle 60°.

DIN ISO 4798 (exept 221-0143 and 221-1181)

 : DIN ISO 4798 (except 221-0143 and 221-1181)

 
  • Funnel O-Ø:
    80 mm
  • Stem O-Ø:
    6 mm
  • Stem length:
    150 mm