Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels with short stem, DURAN®

Funnels with short stem, DURAN®

70 mm, 8 mm, 70 mm
Catalog # 221-1114
Supplier:   DWK Life Sciences
Funnels with short stem, DURAN®
Catalog # 221-1114
Supplier:  DWK Life Sciences
Funnels with short stem, DURAN®
Funnels with short stem, DURAN®

70 mm, 8 mm, 70 mm

Price
19,90 €
Log in for availability
1 * 1 items
Product Details & Documents

DURAN®, borosilicate glass 3.3, clear.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

With smooth internal walls.

DIN ISO 4798 (exept 221-1112/221-1114/221-1120/221-1118 and 221-1119)

Certifications: DIN ISO 4798 (exept 221-1112/221-1114/221-1120/221-1118 and 221-1119).

Documents

Specifications
  • Funnel O-Ø:
    70 mm
  • Stem O-Ø:
    8 mm
  • Stem length:
    70 mm