Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels with short stem, DURAN®

Funnels with short stem, DURAN®

35 mm, 6 mm, 35 mm
Catalog # 221-1111
Supplier:   DWK Life Sciences
Funnels with short stem, DURAN®
Catalog # 221-1111
Supplier:  DWK Life Sciences
Funnels with short stem, DURAN®
Funnels with short stem, DURAN®

35 mm, 6 mm, 35 mm

Price
13,20 €
Log in for availability
1 * 1 items
Product Details & Documents

DURAN®, borosilicate glass 3.3, clear.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

With smooth internal walls.

DIN ISO 4798 (exept 221-1112/221-1114/221-1120/221-1118 and 221-1119)

Certifications: DIN ISO 4798 (exept 221-1112/221-1114/221-1120/221-1118 and 221-1119).

Documents

Specifications
  • Funnel O-Ø:
    35 mm
  • Stem O-Ø:
    6 mm
  • Stem length:
    35 mm