Order Entry
Export
Orders LinkContactUsLinkComponent
Flasks, conical, Erlenmeyer shape, wide neck, DURAN®

Flasks, conical, Erlenmeyer shape, wide neck, DURAN®

3000 ml, 285 mm, 190 mm, 106 mm
Catalog # 201-1902
Supplier:   DWK Life Sciences
Flasks, conical, Erlenmeyer shape, wide neck, DURAN®
Catalog # 201-1902
Supplier:  DWK Life Sciences
Flasks, conical, Erlenmeyer shape, wide neck, DURAN®
Flasks, conical, Erlenmeyer shape, wide neck, DURAN®

3000 ml, 285 mm, 190 mm, 106 mm

Price
108,00 €
Log in for availability
1 * 1 items
Product Details & Documents

DURAN®, borosilicate glass 3.3, clear.

  • Very good chemical resistance, high temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP

With beaded rim and labelling area. Without graduations.

Specifications
  • Ext.Ø:
    190 mm
  • Capacity:
    3000 ml
  • Height:
    285 mm
  • Neck Ø ext.:
    106 mm