Order Entry
Export
Orders LinkContactUsLinkComponent
Test papers, lead acetate papers

Test papers, lead acetate papers

Lead acetate paper for sulfide; Platform: Qualitative test paper, 5 mg/l S²⁻
Catalog # MANA90744
Supplier:   MACHEREY-NAGEL
Test papers, lead acetate papers
Catalog # MANA90744
Supplier:  MACHEREY-NAGEL
Test papers, lead acetate papers
Test papers, lead acetate papers

Lead acetate paper for sulfide; Platform: Qualitative test paper, 5 mg/l S²⁻

Price
15,70 €
Log in for availability
1 * 1 Roll
Product Details & Documents

These test papers allow quick and easy detection of hydrogen sulphide. This gas occurs in the processing of raw oil. It is toxic even in low concentrations. Therefore, critical points are carefully checked. Sulphide containing solutions also give a positive reaction.

  • Colour changes: White to brown-black

Lead acetate is toxic and the test paper needs to be declared as hazardous. The technically equivalent Sulphide test paper is a non-toxic and safe alternative.

Detection limit: Sulfide - 5 mg/L S²⁻

Color change: Sulfide - white → brown-black

Specifications
  • Description:
    Lead acetate paper for sulfide; Platform: Qualitative test paper
  • Seq:
    01
  • Range:
    5 mg/l S²⁻
  • Cat. No.:
    MANA90744
  • Shelf life:
    2 Years
  • Width:
    7 mm