Order Entry
Export
Orders LinkContactUsLinkComponent
 

PEHANON® pH Indicator Test Strips

PEHANON pH 3,8 - 5,5, 200 test strips, Plastic box, 70×15×106 mm, 60 g
Catalog # 662-0215
Supplier:   MACHEREY-NAGEL
PEHANON® pH Indicator Test Strips
Catalog # 662-0215
Supplier:  MACHEREY-NAGEL
PEHANON® pH Indicator Test Strips
PEHANON® pH Indicator Test Strips

PEHANON pH 3,8 - 5,5, 200 test strips, Plastic box, 70×15×106 mm, 60 g

Price
18,60 €
Log in for availability
1 * 200 items
Product Details & Documents

PEHANON® are special pH test strips that unify the pH indicator and the reference colours on one strip. Any sample colour has the same effect on both the reference colours and the reactive pad. Users can be sure to get accurate readings.

  • Sample colour has no effect on the results

  • No accessories needed - always ready to use
  • Highly economic - colour scale on every single strip
  • Safe usage due to hydrophobic barrier

An invisible hydrophobic barrier above the top colour field prevents capillary action of the test solution, so the handle will remain dry and clean. pH values can be determined without a separate colour chart.

 : Not CE certified.

 : No hazardous material.

Specifications
  • Description:
    PEHANON pH 3,8 - 5,5, 200 test strips
  • Seq:
    9
  • Pack type:
    Plastic box, 70×15×106 mm, 60 g
  • Cat. No.:
    662-0215
  • Shelf life:
    3 year(s)