Upright microscope, F1115
Achromatic objective
Catalog # 632-9059
Supplier: MOTIC
Jump to:
Product Details & Documents
The F1115 is designed for applications in education.
- High quality optics with achromatic objectives
- Robust design for students
- Efficient LED energy saving lamp
Delivery information: Supplied with dust cover.
Documents
Specifications
- Magnification:20×/0,40
- Description:Achromatic objective
- Seq:101
- Cat. No.:632-9059
- Nosepiece:Quadruple
- Illumination:LED
Accessories
Product Information
- Catalog #AvailabilityPrice
- Plastic carrying case