Orders LinkContactUsLinkComponent
Caps, LABOCAP, with grip
Caps, LABOCAP, with grip
Catalog # LUDI184110622
Supplier:   LUDI
Caps, LABOCAP, with grip
Catalog # LUDI184110622
Supplier:  LUDI
Caps, LABOCAP, with grip
Caps, LABOCAP, with grip
Price
22,40 €
Log in for availability
1 * 10 items
Product Details & Documents
Aluminium.
  • Made of anodised aluminium with inner stainless steel spring
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
With grip. Cap for sterile but not hermetic closing of test tubes and other vessels.