Caps, LABOCAP, with grip
LABOCAP, silver, 15-16 mm
391-0094
: LUDI
Aluminium.
- Made of anodised aluminium with inner stainless steel spring
- Springs clamp against the outer wall of the vessel
- Anodised and coloured
- Autoclavable
With grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
- For Ø ext.:15-16 mm
- Description:LABOCAP, silver