Upright microscopes, B3 series
Plan achromatic objective
Catalog # 632-8984
Supplier: MOTIC
Jump to:
Product Details & Documents
These microscopes are designed for educational and clinical applications.
- Plan achromatic objectives or:
- φ = Phase contrast
- ASC = Achromatic Super Contrast
Delivery information: Supplied with blue, green and yellow filters, immersion oil and vinyl dust cover.
Specifications
- Magnification:60×/0,85
- Description:Plan achromatic objective
Accessories
Product Information
- Catalog #AvailabilityPrice
- Phase contrast attachment 40×
Product Information
- Catalog #AvailabilityPrice
- Phase contrast attachment 10×