Orders LinkContactUsLinkComponent
Upright microscopes, B3 series
Plan achromatic objective
Catalog # 630-0089
Supplier:   MOTIC
Upright microscopes, B3 series
Catalog # 630-0089
Supplier:  MOTIC
Upright microscopes, B3 series
Upright microscopes, B3 series
Plan achromatic objective
Price
111,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These microscopes are designed for educational and clinical applications.
  • Plan achromatic objectives or:
  • φ = Phase contrast
  • ASC = Achromatic Super Contrast
 : Supplied with blue, green and yellow filters, immersion oil and vinyl dust cover.
Specifications
  • Magnification:
    20×/0,45
  • Description:
    Plan achromatic objective
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Phase contrast attachment 40×
    Phase contrast attachment 40×
    632-8995
    Log in for availability
    1 * 1 items - 86,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Phase contrast attachment 10×
    Phase contrast attachment 10×
    632-8999
    Log in for availability
    1 * 1 items - 86,00 €