Extractor heads for specific light solvents
250 ml
Catalog # LENZ05432149
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, Borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
For the extraction of liquids with specific light solvents, with insert filter (porosity: 0).
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Extraction cap.:250 ml
- Seq:02