Extractors for specific light solvents
Complete set, 100 ml, 250 ml, 45/40 NS
LENZ05432037
DURAN®, Borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
For the extraction of liquids with specific light solvents.
- Extraction cap.:100 ml
- Socket size:45/40 NS
- Type:Complete set
- Flask capacity:250 ml



