Orders LinkContactUsLinkComponent
Adapters, delivery, bent
Adapters, delivery, bent
24/29 NS, 24/29 NS
Catalog # LENZ05265003
Supplier:   Lenz Laborglas
Adapters, delivery, bent
Catalog # LENZ05265003
Supplier:  Lenz Laborglas
Adapters, delivery, bent
Adapters, delivery, bent
24/29 NS, 24/29 NS
Price
33,40 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With core and jacket. Inclined at an angle of 105°. Cone with drip point.
Documents
Specifications
  • Cone size:
    24/29 NS
  • Socket size:
    24/29 NS