Orders LinkContactUsLinkComponent
Adapters, delivery, bent
14/23 NS, 14/23 NS
Catalog # LENZ05265001
Supplier:   Lenz Laborglas
Adapters, delivery, bent
Catalog # LENZ05265001
Supplier:  Lenz Laborglas
Adapters, delivery, bent
Adapters, delivery, bent
14/23 NS, 14/23 NS
Price
19,30 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With core and jacket. Inclined at an angle of 105°. Cone with drip point.
Specifications
  • Cone size:
    14/23 NS
  • Socket size:
    14/23 NS