Order Entry
Export
Orders LinkContactUsLinkComponent
 

Raschig rings

Raschig ring, 8 mm, 8 mm
  LENZ05127008
Raschig rings
  LENZ05127008
Raschig rings
Raschig rings

Raschig ring, 8 mm, 8 mm

 
78,70 €
   
1 * 1 kg
 

Soda lime glass.

  • For packing columns
  • Good chemical resistance
  • Good thermal resistance
 
  • Ext.Ø:
    8 mm
  • Description:
    Raschig ring
  • Height:
    8 mm
  • Seq:
    05