Orders LinkContactUsLinkComponent
Distilling bridges
Distilling bridges
300 mm, 24/29 NS
Catalog # 201-0347
Supplier:   Lenz Laborglas
Distilling bridges
Catalog # 201-0347
Supplier:  Lenz Laborglas
Distilling bridges
Distilling bridges
300 mm, 24/29 NS
Price
60,60 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Still heads without condenser, with two vertical cones.
Documents
Specifications
  • Cone size:
    24/29 NS
  • Jacket length:
    300 mm