Distilling bridges
200 mm, 19/26 NS
201-0346
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Still heads without condenser, with two vertical cones.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
- Cone size:19/26 NS
- Jacket length:200 mm