Orders LinkContactUsLinkComponent
Splash heads, Stutzer, bent
Splash heads, Stutzer, bent
24/29 NS
Catalog # 201-0344
Supplier:   Lenz Laborglas
Splash heads, Stutzer, bent
Catalog # 201-0344
Supplier:  Lenz Laborglas
Splash heads, Stutzer, bent
Splash heads, Stutzer, bent
24/29 NS
Price
81,80 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Two standard ground cones, inclined at an angle of 75°. Placed directly on the distillation flask to intercept droplets.
Documents
Specifications
  • Cone size:
    24/29 NS