Adapters, distilling heads
14/23 NS, 19/26 - 19/26 NS
Catalog # 201-0339
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Includes a socket for a distillation thermometer of 55 mm fitted length and two cones at an angle of 75°.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Cone size:19/26 - 19/26 NS
- Socket size:14/23 NS