Adapters, distilling heads
14/23 NS, 14/23 - 14/23 NS
201-0338
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Includes a socket for a distillation thermometer of 55 mm fitted length and two cones at an angle of 75°.
- Cone size:14/23 - 14/23 NS
- Socket size:14/23 NS