Orders LinkContactUsLinkComponent
Dropping funnels, conical shape, without pressure balance
Dropping funnels, conical shape, without pressure balance
100 ml, 19/26 NS, 19/26 NS
Catalog # 201-0910
Supplier:   Lenz Laborglas
Dropping funnels, conical shape, without pressure balance
Catalog # 201-0910
Supplier:  Lenz Laborglas
Dropping funnels, conical shape, without pressure balance
Dropping funnels, conical shape, without pressure balance
100 ml, 19/26 NS, 19/26 NS
Price
69,10 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3.3.
  • Screw thread retaining device
  • With inscription label, without graduation
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and stopcock with glass or PTFE plug.
Documents
Specifications
  • Cone size:
    19/26 NS
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml