Flasks, flat bottom with standard ground joint
100 ml, 110 mm, 64 mm, 29/32 NS
201-1202
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
DIN 12348
Certifications: DIN 12348
- Ext.Ø:64 mm
- Socket size:29/32 NS
- Capacity:100 ml
- Height:110 mm
- Seq:06



