Flasks, flat bottom with standard ground joint
500 ml, 165 mm, 105 mm, 19/26 NS
201-1214
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
DIN 12348
: DIN 12348
- Ext.Ø:105 mm
- Socket size:19/26 NS
- Capacity:500 ml
- Height:165 mm
- Seq:10



