Flasks, flat bottom with standard ground joint
50 ml, 100 mm, 51 mm, 19/26 NS
201-1211
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
DIN 12348
: DIN 12348
- Ext.Ø:51 mm
- Socket size:19/26 NS
- Capacity:50 ml
- Height:100 mm
- Seq:01



