Microtube racks, reversible
Hi-vis orange, 96, 0,2/0,5 ml, Reversible
Catalog # 700-2825
Supplier: Thermo Fisher Scientific
Microtube racks, reversible
Catalog # 700-2825
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Made of PP.
- Excellent chemical resistance
- Excellent temperature resistance
- Sturdy
These racks are for microcentrifuge tubes with molded alphanumeric grid and also assorted colours are available.
Specifications
- For tubes:0,2/0,5 ml
- Form:Reversible
- No. of Holes:96
- Colour:Hi-vis orange
- Cat. No.:700-2825
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact