Orders LinkContactUsLinkComponent
Aerosol analysis monitors
Absorbent pad, 25 mm
  MILFAP1002500
 :   Merck
Aerosol analysis monitors
  MILFAP1002500
 :  Merck
Aerosol analysis monitors
Aerosol analysis monitors
Absorbent pad, 25 mm
60,90 €
1 * 100 items
The PS monitor is used for sample collection and particle analysis. The PP monitor is used for microscopic particle analysis and sample collection for HPLC and UV detection.
Membrane: mixed cellulose esters with cellulose support pad; housing: PS or carbon filled PP.
  • Description:
    Absorbent pad, 25 mm