Order Entry
Export
Orders LinkContactUsLinkComponent
Pipette tips, Finntip™ Stepper

Pipette tips, Finntip™ Stepper

1,25 ml, +, Individually
Catalog # 613-2637
   Low Manufacturing Impact
Pipette tips, Finntip™ Stepper
Catalog # 613-2637
   Low Manufacturing Impact
Pipette tips, Finntip™ Stepper
Pipette tips, Finntip™ Stepper

1,25 ml, +, Individually

Price
133,00 €
Log in for availability
1 * 50 items
Product Details & Documents

Full range of positive displacement syringe tips designed for the Finnpipette™ Stepper.

  • Ergonomic design
  • Seven syringe sizes covering a range of dispensing volumes from 10 µl to 50 ml
  • Adapted for applications using aggressive and viscous liquids
Specifications
  • Packed:
    Individually
  • Sterile:
    +
  • Volume:
    1,25 ml
  • Seq:
    04
  • Cat. No.:
    613-2637
  • Environmental certification:
    ISO 14001 (Certificate Registration No. EMS 623400)
  • Environmentally Preferable:
    Low Manufacturing Impact