Bottles, narrow neck, square, HDPE, with thread
500 ml, 32 mm, 74×74×170 mm
Catalog # 215-2368
Supplier: Kautex
Jump to:
Product Details & Documents
HDPE. Square. Natural or white colour.
- Square cross-section
- Recessed labelling field
- Leakproof
- Bottles are suitable for food
- Ideal for liquids
- Halal certified
Five different types of closures available.
Documents
Specifications
- Thread:32 mm
- Capacity:500 ml
- Seq:03
- W×D×H:74×74×170 mm



