Drawer organiser
5, 400×300×65 mm
Catalog # 216-6420
Supplier: KARTELL
Jump to:
Product Details & Documents
PVC, white.
- Version with 12 compartments for small items, e.g. magnetic stirrer bars
- Version with 5 compartments for small parts of various sizes, e.g. pipettes, thermometers, test tubes, spatulas, more
- Smooth surfaces with rounded corners for easy cleaning
For well organised storage of many items in drawers, on shelves or at the workstation.
Specifications
- Compartments:5
- Seq:01
- W×D×H:400×300×65 mm