Orders LinkContactUsLinkComponent
Drawer organiser
12, 400×300×65 mm
  216-6421
 :   KARTELL
Drawer organiser
  216-6421
 :  KARTELL
Drawer organiser
Drawer organiser
12, 400×300×65 mm
83,50 €
1 * 1 items
PVC, white.
  • Version with 12 compartments for small items, e.g. magnetic stirrer bars
  • Version with 5 compartments for small parts of various sizes, e.g. pipettes, thermometers, test tubes, spatulas, more
  • Smooth surfaces with rounded corners for easy cleaning
For well organised storage of many items in drawers, on shelves or at the workstation.
  • Compartments:
    12
  • Seq:
    02
  • W×D×H:
    400×300×65 mm