Orders LinkContactUsLinkComponent
Desiccator plates
Desiccator plates
PP, 238 mm
Catalog # 467-2112
Supplier:   KARTELL
Desiccator plates
Catalog # 467-2112
Supplier:  KARTELL
Desiccator plates
Desiccator plates
PP, 238 mm
Price
25,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Perforated PP desiccator plates. Suitable for use with desiccators: 467-2115, 467-2120 and 467-2125. Use at room temperature only, not suitable for use with hot crucibles.
Specifications
  • Ø:
    238 mm
  • Type:
    PP
  • Seq:
    03