Desiccator plates
PP, 141 mm
Catalog # 467-2110
Supplier: KARTELL
Jump to:
Product Details & Documents
Perforated PP desiccator plates. Suitable for use with desiccators: 467-2115, 467-2120 and 467-2125. Use at room temperature only, not suitable for use with hot crucibles.
Specifications
- Ø:141 mm
- Type:PP
- Seq:01