Orders LinkContactUsLinkComponent
Drying tubes, calcium chloride
8-10 mm, 270 mm, 20x200 mm
Catalog # 468-1212
Supplier:   KARTELL
Drying tubes, calcium chloride
Catalog # 468-1212
Supplier:  KARTELL
Drying tubes, calcium chloride
Drying tubes, calcium chloride
8-10 mm, 270 mm, 20x200 mm
Price
6,20 €
Log in for availability
1 * 1 items
Product Details & Documents
PE.
  • Both ends have tapered, push-on connectors
Suitable for drying gases.
Specifications
  • Ø×L:
    20x200 mm
  • For tube Ø:
    8-10 mm
  • Total length:
    270 mm