Drying tubes, calcium chloride
8-10 mm, 220 mm, 20x150 mm
Catalog # 468-1211
Supplier: KARTELL
Jump to:
Product Details & Documents
PE.
- Both ends have tapered, push-on connectors
Suitable for drying gases.
Specifications
- Ø×L:20x150 mm
- For tube Ø:8-10 mm
- Total length:220 mm