Orders LinkContactUsLinkComponent
Desiccator plates
Desiccator plates
PP, 189 mm
Catalog # 467-2111
Supplier:   KARTELL
Desiccator plates
Catalog # 467-2111
Supplier:  KARTELL
Desiccator plates
Desiccator plates
PP, 189 mm
Price
16,20 €
Log in for availability
1 * 1 items
Product Details & Documents
Perforated PP desiccator plates. Suitable for use with desiccators: 467-2115, 467-2120 and 467-2125. Use at room temperature only, not suitable for use with hot crucibles.
Specifications
  • Ø:
    189 mm
  • Type:
    PP
  • Seq:
    02