Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

230 mm, 220 mm
Catalog # 221-1525
Supplier:   KARTELL
Funnels with short stem
Catalog # 221-1525
Supplier:  KARTELL
Funnels with short stem
Funnels with short stem

230 mm, 220 mm

Price
15,50 €
Log in for availability
1 * 1 items
Product Details & Documents

HDPE, translucent.

  • Good chemical resistance
  • With raised lip to prevent overflowing
  • External ribs for unobstructed removal of air when filtering

With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.

Documents

Specifications
  • Funnel O-Ø:
    220 mm
  • Length:
    230 mm
  • Seq:
    06