Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bottles, wide neck, with screw cap

250 ml, GL 45, 38 mm, 140 mm, 60 mm
  215-0906
 :   KARTELL
Bottles, wide neck, with screw cap
  215-0906
 :  KARTELL
Bottles, wide neck, with screw cap
Bottles, wide neck, with screw cap

250 ml, GL 45, 38 mm, 140 mm, 60 mm

 
7,60 €
   
1 * 1 items
 

PP, translucent with PP screw cap.

  • Good temperature and chemical resistance
  • Compliant with food and drug regulations

Sturdy graduated bottle with a long pinch seal that ensures leakproof closure. Wide neck for easy filling and emptying of liquids and powders.

DIN 13316 and 168

 : DIN 13316 and 168

 
  • Ext.Ø:
    60 mm
  • Thread:
    GL 45
  • Capacity:
    250 ml
  • Height:
    140 mm
  • Neck I-Ø:
    38 mm