Stoppers for conical ground joints
Ribbed cap, Black, 29/32, 25,8 mm
612-0947
: KARTELL
Stoppers with coloured insert in the holding disc for better identification.
- Transparent LDPE
- Good chemical resistance
- Sealing rings for impermeable and secure fit
Pentagonal holding disc for easy loosening and removal of the stopper.
- Ø bottom:25,8 mm
- NS:29/32
- Colour:Black
- Description:Ribbed cap