Stoppers for conical ground joints
Ribbed cap, Yellow, 19/26, 16,4 mm
Catalog # 612-0946
Supplier: KARTELL
Jump to:
Product Details & Documents
Stoppers with coloured insert in the holding disc for better identification.
- Transparent LDPE
- Good chemical resistance
- Sealing rings for impermeable and secure fit
Pentagonal holding disc for easy loosening and removal of the stopper.
Specifications
- Ø bottom:16,4 mm
- NS:19/26
- Colour:Yellow
- Description:Ribbed cap