Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® Buffer Tablets for Haematology

VWR® Buffer Tablets for Haematology

Buffer tablets, 7,2, 100 ml, 50 Tab.
Catalog # 331942F
Supplier:   VWR Chemicals
VWR® Buffer Tablets for Haematology
Catalog # 331942F
Supplier:  VWR Chemicals
CAS Number:  
VWR® Buffer Tablets for Haematology
VWR® Buffer Tablets for Haematology

Buffer tablets, 7,2, 100 ml, 50 Tab.

CAS Number:  
Price
53,40 €
Log in for availability
1 * 50 Tablet
Product Details & Documents

These tablets provide a rapid and convenient method of preparing a buffer solution.

Gurr buffer tablets for the preparation of microscopical stain solutions.

One tablet dissolved in distilled water, and made up to 100 ml, produces a solution of the stated pH at 20 °C. The molarity of the buffer solutions is 0,005 M.

Specifications

Specifications

  • Pk:
    50 Tab.
  • Description:
    Buffer tablets
  • Final volume:
    100 ml
  • pH:
    7,2

Specifications